Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, Thermo Scientific Chemicals
Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, Thermo Scientific Chemicals
Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, Thermo Scientific Chemicals
Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Catalog number ACR423175000
Price (MYR)
542.00
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
542.00
Add to cart
Chemical Identifiers
CAS17372-87-1
IUPAC Namedisodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6Br4Na2O5
InChI KeySEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<10 %
Specific extinction E (1%/1cm)on dried substance
Appearance (Form)Crystalline powder
Specific extinction E (1%/1cm)>=1100 (lambda max, 0.001M Na2CO3)
View more
For UV/VIS spectroscopy
RUO – Research Use Only