Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number ACR423152500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS142-63-2
IUPAC Namepiperazine hexahydrate
Molecular FormulaC4H22N2O6
InChI KeyAVRVZRUEXIEGMP-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.C1CNCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Infrared spectrumConforms
Titration with HCl>=97.5 %
Appearance (Color)Transparent to white