Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR423150051
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS525-82-6
IUPAC Name2-phenyl-4H-chromen-4-one
Molecular FormulaC15H10O2
InChI KeyVHBFFQKBGNRLFZ-UHFFFAOYSA-N
SMILESO=C1C=C(OC2=CC=CC=C12)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)95-98?C
Appearance (Color)White to pale cream
Assay (GC)≥98.5%