Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
Plastic bottle
Catalog number ACR423150010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOxalic acid dihydrate
Name NoteFor analysis ACS
CAS6153-56-6
Health Hazard 1Danger