Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ACR422750250
Price (MYR)
1,467.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,467.00
Add to cart
Chemical Identifiers
CAS89-51-0
IUPAC Name2-(carboxymethyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyZHQLTKAVLJKSKR-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
Appearance (Color)White to cream to pale yellow