Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR422750050
Price (MYR)
578.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
578.00
Add to cart
Chemical Identifiers
CAS207596-32-5
IUPAC Namepalladium(2+) dinitrate
Molecular FormulaN2O6Pd
InChI KeyGPNDARIEYHPYAY-UHFFFAOYSA-N
SMILES[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline
Appearance (Color)Reddish brown
Assay (special)Pd 39.0-39.9%
Total Metal Impurities≤0.2%