pH Indicator solution, universal (in isopropanol), pure
pH Indicator solution, universal (in isopropanol), pure
Thermo Scientific Chemicals
pH Indicator solution, universal (in isopropanol), pure
Have Questions?
Catalog NumberQuantity
ACR422435000500 mL
Catalog number ACR422435000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS269409-99-6
IUPAC Nameethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular FormulaC15H21BO4
InChI KeyRFBZWPFBCXBBJS-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=C1B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)56.0-66.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to yellow
Assay (GC)≥96.0%
Indicator
RUO – Research Use Only