Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
CAS: 14634-91-4 | C36H24FeN6O4S | 692.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR422251000
Price (MYR)
656.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
656.00
Add to cart
Chemical Identifiers
CAS3304-51-6
IUPAC Name2-amino-5-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular FormulaC13H18N2O4
InChI KeyVULSXQYFUHKBAN-UHFFFAOYNA-N
SMILESNC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Indicator
RUO – Research Use Only