2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number ACR421985000
Price (MYR)
501.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
501.00
Add to cart
Chemical Identifiers
CAS58917-26-3
IUPAC Name6-methylhept-5-en-2-ol
Molecular FormulaC8H16O
InChI KeyOHEFFKYYKJVVOX-UHFFFAOYNA-N
SMILESCC(O)CCC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4470-1.4500 @ 20?C
FormLiquid
Optical Rotation18.1 ± 0.1? (neat)
For UV/VIS spectroscopy
RUO – Research Use Only