1,1,2-Trimethyl-1H-benz[e]indole, 96%
1,1,2-Trimethyl-1H-benz[e]indole, 96%
1,1,2-Trimethyl-1H-benz[e]indole, 96%
1,1,2-Trimethyl-1H-benz[e]indole, 96%
Thermo Scientific Chemicals
1,1,2-Trimethyl-1H-benz[e]indole, 96%
CAS: 41532-84-7 | C15H15N | 209.29 g/mol
Have Questions?
Catalog number ACR421900250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS437-83-2
IUPAC Name3-fluoro-2-methoxyaniline
Molecular FormulaC7H8FNO
InChI KeyRCYMPYMITUEHOJ-UHFFFAOYSA-N
SMILESCOC1=C(N)C=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5310-1.5360 @ 20?C
Appearance (Color)Clear orange