Trilaurin, 98%, Thermo Scientific Chemicals
Trilaurin, 98%, Thermo Scientific Chemicals
Trilaurin, 98%, Thermo Scientific Chemicals
Trilaurin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trilaurin, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR421830050
Price (MYR)
576.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
576.00
Add to cart
Specifications
Chemical Name or MaterialTrilaurin
CAS538-24-9
Melting Point45°C to 47°C
ColorWhite
Density0.9000g/mL