Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals
Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals
Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals
Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals
CAS: 2622-14-2 | C18H33P | 280.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR421610250
Price (MYR)
2,537.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,537.00
Add to cart
Chemical Identifiers
CAS122-04-3
IUPAC Name4-nitrobenzoyl chloride
Molecular FormulaC7H4ClNO3
InChI KeySKDHHIUENRGTHK-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystalline powder or crystals or flakes
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)70-75?C