Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
CAS: 87-90-1 | C3Cl3N3O3 | 232.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
Catalog number ACR421542500
Price (MYR)
496.00
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
496.00
Add to cart
Chemical Identifiers
CAS14691-89-5
IUPAC Name(4-acetamido-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Molecular FormulaC11H21N2O2
InChI KeyUXBLSWOMIHTQPH-UHFFFAOYSA-N
SMILESCC(=O)NC1CC(C)(C)N([O])C(C)(C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Orange to red
FormCrystals or powder or crystalline powder
C-X bond formation reagent
RUO – Research Use Only