Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
CAS: 87-90-1 | C3Cl3N3O3 | 232.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass Bottle
Catalog number ACR421540010
Price (MYR)
690.00
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
690.00
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Platinum
FormFoil
Total Metal Impurities≤0.1%
C-X bond formation reagent
RUO – Research Use Only