Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass Bottle
Catalog number ACR421455000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS112-38-9
IUPAC Nameundec-10-enoic acid
Molecular FormulaC11H20O2
InChI KeyFRPZMMHWLSIFAZ-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale yellow
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormFused solid or clear liquid as melt
Assay (GC)≥97.0%