Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Catalog number ACR421451000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTrichloroacetic acid
Name Note99+%
CAS76-03-9
Health Hazard 1Danger