2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
CAS: 75-80-9 | C2H3Br3O | 282.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number ACR421430500
Price (MYR)
1,343.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,343.00
Add to cart
Chemical Identifiers
CAS81-96-9
IUPAC Name14-bromotetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(17),2,4,6,9,11,13,15-octaen-8-one
Molecular FormulaC17H9BrO
InChI KeyWVECFEIAZAKUNF-UHFFFAOYSA-N
SMILESBrC1=C2C=CC=C3C(=O)C4=CC=CC=C4C(C=C1)=C23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale green to green to yellow to brown
Assay (HPLC)≥80.0%
FormPowder
Melting Point (clear melt)164.5-173.5?C