2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
CAS: 75-80-9 | C2H3Br3O | 282.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR421430100
Price (MYR)
573.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
573.00
Add to cart
Chemical Identifiers
CAS122-39-4
IUPAC NameN-phenylaniline
Molecular FormulaC12H11N
InChI KeyDMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
Melting point52.5°C to 54.0°C
Residue after ignition=<0.03 %