p-Toluenesulfonic acid, sodium salt, Thermo Scientific Chemicals
p-Toluenesulfonic acid, sodium salt, Thermo Scientific Chemicals
p-Toluenesulfonic acid, sodium salt, Thermo Scientific Chemicals
p-Toluenesulfonic acid, sodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid, sodium salt, Thermo Scientific Chemicals
CAS: 657-84-1 | C7H7NaO3S | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR421231000
Price (MYR)
574.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
574.00
Add to cart
Chemical Identifiers
CAS2695-37-6
IUPAC Namesodium 4-ethenylbenzene-1-sulfonate
Molecular FormulaC8H7NaO3S
InChI KeyXFTALRAZSCGSKN-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Sulfated Ash30.31-38.57% (Dry wt basis)
FormPowder
Water Content (Karl Fischer Titration)5.0 to 13.6% (0.6 to 1.8 waters)
Assay (ex Sulphated Ash)≥88.0 to ≤112.0% (Dry wt basis)