p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Catalog number ACR421215000
Price (MYR)
814.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
814.00
Add to cart
Chemical Identifiers
CAS372-75-8
IUPAC Name2-amino-5-(carbamoylamino)pentanoic acid
Molecular FormulaC6H13N3O3
InChI KeyRHGKLRLOHDJJDR-UHFFFAOYNA-N
SMILESNC(CCCNC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.3 % (1 g, 105°C, 3 hrs)
UV transmittance>=95.0 % (c=10, H2O, 430 nm) 1 cm cell
Ammonium (NH4)=<200 ppm
Iron (Fe)=<20 ppm