p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number ACR421210250
Price (MYR)
362.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
362.00
Add to cart
Chemical Identifiers
CAS12125-02-9
IUPAC Nameammonium chloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES[NH4+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormGranular
Appearance (Color)White to pale yellow
Assay (unspecified)99.5% min