o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
CAS: 612-82-8 | C14H16N2·2HCl | 285.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass Bottle
Catalog number ACR421131000
Price (MYR)
1,133.00
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,133.00
Add to cart
Chemical Identifiers
CAS1003-03-8
IUPAC Namecyclopentanamine
Molecular FormulaC5H11N
InChI KeyNISGSNTVMOOSJQ-UHFFFAOYSA-N
SMILESNC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.3 %
Appearance (Form)Clear liquid
GC>=99.0 %
Refractive index1.4482 to 1.4502 (20°C, 589 nm)