DL-α-Tocopheryl acetate, 98%
DL-α-Tocopheryl acetate, 98%
DL-α-Tocopheryl acetate, 98%
DL-α-Tocopheryl acetate, 98%
Thermo Scientific Chemicals
DL-α-Tocopheryl acetate, 98%
CAS: 7695-91-2 | C31H52O3 | 472.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR421070010
Price (MYR)
411.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
411.00
Add to cart
Chemical Identifiers
CAS2437-56-1
IUPAC Nametridec-1-ene
Molecular FormulaC13H26
InChI KeyVQOXUMQBYILCKR-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4315-1.4365 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%