D-δ-Tocopherol, 93%, Thermo Scientific Chemicals
D-δ-Tocopherol, 93%, Thermo Scientific Chemicals
D-δ-Tocopherol, 93%, Thermo Scientific Chemicals
D-δ-Tocopherol, 93%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D-δ-Tocopherol, 93%, Thermo Scientific Chemicals
CAS: 119-13-1 | C27H46O2 | 402.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number ACR421051000
Price (MYR)
1,239.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,239.00
Add to cart
Chemical Identifiers
CAS403-19-0
IUPAC Name2-fluoro-4-nitrobenzen-1-olate
Molecular FormulaC6H3FNO3
InChI KeyORPHLVJBJOCHBR-UHFFFAOYSA-M
SMILES[O-]C1=CC=C(C=C1F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to pale brown
Melting Point (clear melt)115.0-122.0?C
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.0%