D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
Thermo Scientific Chemicals
D-gamma-Tocopherol, 95%
CAS: 54-28-4 | C28H48O2 | 416.69 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR421041000100 mgGlass bottle
Catalog number ACR421041000
Price (MYR)
1,668.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,668.00
Add to cart
Specifications
Chemical Name or MaterialD-γ-Tocopherol
Name Note95%
CAS54-28-4
ColorBrown
EINECS Number200-201-5