D-alpha-Tocopherol, 97+%
D-alpha-Tocopherol, 97+%
D-alpha-Tocopherol, 97+%
D-alpha-Tocopherol, 97+%
Thermo Scientific Chemicals
D-alpha-Tocopherol, 97+%
CAS: 59-02-9 | C29H50O2 | 430.7 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR421031000100 mgGlass Bottle
Catalog number ACR421031000
Price (MYR)
884.00
Add to cart
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
884.00
Add to cart
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Appearance (Color)Gray
Comment2.5 cm and down
FormSolid