Secador™ Standard Desiccator Cabinet, Vertical Model
Secador™ Standard Desiccator Cabinet, Vertical Model
Actual product may vary
Secador™ Standard Desiccator Cabinet, Vertical Model
Secador™ Standard Desiccator Cabinet, Vertical Model
Thermo Scientific Chemicals
Secador™ Standard Desiccator Cabinet, Vertical Model
Constructed of Durastar™ co-polyester. Bel-Art™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing, and chemical attack.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of ShelvesVolume (English)Height (Metric)
ACR420731000Clear31.58 cu. ft.41.7 cm
FIS08-647-657Clear31.58 cu. ft.41.7 cm
FKS12318995Clear31.58 cu. ft.41.7 cm
FSA420731000Clear31.58 cu. ft.41.7 cm
ACR420721000Clear21.17 cu. ft.31.4 cm
FSA420721000Clear21.17 cu. ft.31.4 cm
Catalog number ACR420731000
Price (MYR)
1,213.00
Add to cart
Color:
Clear
No. of Shelves:
3
Volume (English):
1.58 cu. ft.
Height (Metric):
41.7 cm
Price (MYR)
1,213.00
Add to cart
Constructed of Durastar™ co-polyester. Bel-Art™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing, and chemical attack.
  • Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Compact 34.1 W x 41.4cm D footprint for space saving stacking up to 3 units high
  • Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
  • Can be used with desiccant cartridges, F42049-0100 or F42046-0000
Specifications
AutoclavableNot Autoclavable
ColorClear
Cabinet StyleVertical
MaterialCopolyester
Product TypeDesiccator
Door StyleLeft-hinged and gasketed
For Use With (Application)Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant
IncludesCarrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model3.0
No. of Shelves3
ResistanceChemicals, stains and fine cracks
Shelf TypeRemovable Perforated
Volume (English)1.58 cu. ft.
Dimensions (L x W)16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English)16.3 in.
Length (Metric)41.4 cm
Width (English)13.4 in.
Width (English) Door8.9 in.
Width (Metric)34.1 cm
Width (Metric) Door22.6 cm
Height (English)16.4 in.
Height (English) Opening12.3 in.
Height (Metric)41.7 cm
Depth (English)16.3 in.
Depth (Metric)41.4 cm
Unit Size