Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
CAS: 2537-36-2 | C4H12ClNO4 | 173.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR420561000
Price (MYR)
2,883.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,883.00
Add to cart
Specifications
Chemical Name or MaterialTetramethylammonium perchlorate
CAS2537-36-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May intensify fire; oxidizer.
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Oxidation reagent, Phase transfer catalyst
RUO – Research Use Only