L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
CAS: 87-69-4 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number ACR420005000
Price (MYR)
759.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
759.00
Add to cart
Specifications
Chemical Name or MaterialL(+)-Tartaric acid
Name Note99+%
CAS87-69-4
Health Hazard 1Danger