Stoddard Solvent, Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
CAS: 8052-41-3 | C8H15BrO2 | 223.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number ACR419730050
Price (MYR)
2,462.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
2,462.00
Add to cart
Chemical Identifiers
CAS586-98-1
IUPAC Name(pyridin-2-yl)methanol
Molecular FormulaC6H7NO
InChI KeySHNUBALDGXWUJI-UHFFFAOYSA-N
SMILESOCC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Infrared spectrumConforms
Refractive index1.5410 to 1.5450 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid