Stearic anhydride, 98%
Stearic anhydride, 98%
Stearic anhydride, 98%
Stearic anhydride, 98%
Thermo Scientific Chemicals
Stearic anhydride, 98%
CAS: 638-08-4 | C36H70O3 | 550.95 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR41971025025 gGlass bottle
Catalog number ACR419710250
Price (MYR)
1,654.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,654.00
Add to cart
Chemical Identifiers
CAS51-17-2
IUPAC Name1H-1,3-benzodiazole
Molecular FormulaC7H6N2
InChI KeyHYZJCKYKOHLVJF-UHFFFAOYSA-N
SMILESN1C=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point171°C to 174°C
HPLC>=97.5 %
Appearance (Color)Beige to brown
Appearance (Form)Crystalline powder or crystals