Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number ACR419330250
Price (MYR)
3,185.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,185.00
Add to cart
Chemical Identifiers
CAS1921-70-6
IUPAC Name2,6,10,14-tetramethylpentadecane
Molecular FormulaC19H40
InChI KeyXOJVVFBFDXDTEG-UHFFFAOYNA-N
SMILESCC(C)CCCC(C)CCCC(C)CCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid