Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number ACR419330100
Price (MYR)
1,477.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,477.00
Add to cart
Chemical Identifiers
CAS306-94-5
IUPAC Nameoctadecafluoro-decahydronaphthalene
Molecular FormulaC10F18
InChI KeyUWEYRJFJVCLAGH-UHFFFAOYSA-N
SMILESFC1(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C1(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=88 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless