Rosin, gum, Thermo Scientific Chemicals
Rosin, gum, Thermo Scientific Chemicals
Rosin, gum, Thermo Scientific Chemicals
Rosin, gum, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Rosin, gum, Thermo Scientific Chemicals
CAS: 8050-09-7 | C20H30O2 | 302.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Catalog number ACR419170025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS105-54-4
IUPAC Nameethyl butanoate
Molecular FormulaC6H12O2
InChI KeyOBNCKNCVKJNDBV-UHFFFAOYSA-N
SMILESCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.3910-1.3940 @ 20?C
FormLiquid
CommentClear colorless
Adsorbent
RUO – Research Use Only