Pyridine, 99+%, extra pure, nonaqueous titration grade, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, nonaqueous titration grade, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, nonaqueous titration grade, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, nonaqueous titration grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, nonaqueous titration grade, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass Bottle
Catalog number ACR418530010
Price (MYR)
865.00
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
865.00
Add to cart
Chemical Identifiers
CAS665-66-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)<0.5%
FormPowder or crystalline powder