2-Propene-1-sulfonic acid sodium salt, 95%, pract.
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
Thermo Scientific Chemicals
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
CAS: 2495-39-8 | C3H5NaO3S | 144.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR41837025025 gGlass bottle
Catalog number ACR418370250
Price (MYR)
777.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
777.00
Add to cart
Chemical Identifiers
CAS7487-94-7
IUPAC Namemercury(2+) dichloride
Molecular FormulaCl2Hg
InChI KeyLWJROJCJINYWOX-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White