Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR417890250
Price (MYR)
738.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
738.00
Add to cart
Chemical Identifiers
CAS69833-18-7
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Oxidation reagent
RUO – Research Use Only