p-Phenylene diisothiocyanate, 99%, Thermo Scientific Chemicals
p-Phenylene diisothiocyanate, 99%, Thermo Scientific Chemicals
p-Phenylene diisothiocyanate, 99%, Thermo Scientific Chemicals
p-Phenylene diisothiocyanate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Phenylene diisothiocyanate, 99%, Thermo Scientific Chemicals
CAS: 4044-65-9 | C8H4N2S2 | 192.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4175100505 gGlass bottle
Catalog number ACR417510050
Price (MYR)
788.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
788.00
Add to cart
Chemical Identifiers
CAS67952-60-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Infrared spectrumConforms
Refractive index1.5190 to 1.5230 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=94 %