Petrolatum, white, pure, Thermo Scientific Chemicals
Petrolatum, white, pure, Thermo Scientific Chemicals
Petrolatum, white, pure, Thermo Scientific Chemicals
Petrolatum, white, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petrolatum, white, pure, Thermo Scientific Chemicals
CAS: 8009-03-8 | C15H15N | 209.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number ACR417090010
Price (MYR)
475.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
475.00
Add to cart
Specifications
Chemical Name or MaterialPetrolatum, white
CAS8009-03-8
Health Hazard 2GHS H Statement:
Health Hazard 3GHS P Statement:
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Melting Point50.0°C to 56.0°C