Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
CAS: 422-64-0 | C3HF5O2 | 164.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Plastic bottle
Catalog number ACR416920500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS69739-34-0
IUPAC Nametert-butyldimethylsilyl trifluoromethanesulfonate
Molecular FormulaC7H15F3O3SSi
InChI KeyWLLIXJBWWFGEHT-UHFFFAOYSA-N
SMILESCC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3835-1.3880 @ 20?C
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%