Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, CH2O, CAS Number-30525-89-4, paraformaldehyde, paraform, oxymethylene, methylene oxide, methyl aldehyde, methanal, formol, formalin, formic aldehyde, oxomethane, 500g, CHEBI:16842, White, 260 deg.C, 1.03 (Air - 1.0), 71 deg.C, Extra Pure, Warning
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number ACR416785000
Price (MYR)
525.00
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
525.00
Add to cart
Chemical Identifiers
CAS10025-77-1
IUPAC Nameiron(3+) hexahydrate trichloride
Molecular FormulaCl3FeH12O6
InChI KeyNQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to light brown
Appearance (Form)Adhering powder and/or lumps
Titration Iodometric97.0 to 102.0 %
Insoluble matter=<0.01 %
Phosphorous compounds (as PO4)=<0.01 %