Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, CH2O, CAS Number-30525-89-4, paraformaldehyde, paraform, oxymethylene, methylene oxide, methyl aldehyde, methanal, formol, formalin, formic aldehyde, oxomethane, 500g, CHEBI:16842, White, 260 deg.C, 1.03 (Air - 1.0), 71 deg.C, Extra Pure, Warning
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR416780010
Price (MYR)
646.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
646.00
Add to cart
Chemical Identifiers
CAS25322-68-3
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Solution TestClear, APHA ≤30
Viscosity550 - 750 mPas (50%, water @ 20?C)
Appearance (Color)White
FormPowder or crystals or flakes
CommentAveraged mol weight 9000 - 11250 (based on OH value)