Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
2 kg
Catalog number ACR416770100
Price (MYR)
2,663.00
Add to cart
Quantity:
10 kg
Request bulk or custom format
Price (MYR)
2,663.00
Add to cart
Specifications
Chemical Name or MaterialParaffin wax
CAS8002-74-2
Melting Point58.0°C to 62.0°C
ColorWhite
RTECS NumberRV0350000
View more
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
RUO – Research Use Only