Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
10 kg
Catalog number ACR416770020
Price (MYR)
855.00
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
855.00
Add to cart
Chemical Identifiers
CAS108-10-1
IUPAC Name4-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyNTIZESTWPVYFNL-UHFFFAOYSA-N
SMILESCC(C)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.3950 to 1.3970 (20°C, 589 nm)
Calcium (Ca)=<0.5 ppm
Chromium (Cr)=<0.02 ppm
Color scale=<15 APHA
View more
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
RUO – Research Use Only