Pancreatin, for biochemistry, powder, CAS 8049-47-6
Pancreatin, for biochemistry, powder, CAS 8049-47-6
Thermo Scientific Chemicals
Pancreatin, for biochemistry, powder, CAS 8049-47-6
Pancreatin, CAS 8049-47-6, is an extract of animal pancreas that is used to digest polymers. | CAS: 8049-47-6
Have Questions?
Catalog NumberQuantity
ACR416745000
Catalog number ACR416745000
Price (MYR)
1,738.00
Add to cart
Request bulk or custom format
Price (MYR)
1,738.00
Add to cart
Chemical Identifiers
CAS7789-00-6
IUPAC Namedipotassium dioxochromiumbis(olate)
Molecular FormulaCrK2O4
InChI KeyXMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric>=99.5 %
Iron (Fe)=<20 ppm
Appearance (Form)Crystals
Calcium (Ca)=<50 ppm
Chloride (Cl)=<10 ppm
View more
General Description
  • Pancreatin is an extract that contains the enzymes amylase, trypsin, and lipase
  • CAS 8049-47-6
Applications
  • It can be used to digest protein, lipid, and starch in solutions
  • Useful for digestion of culture cell adhesions, extracellular matrix components, stored starches and lipids, and more in vitro, or for biochemistry applications
RUO – Research Use Only
Literature: Enna, S.J.; Bylund, David B. Pancreatin. xPharm: The Comprehensive Pharmacology Reference,Elsevier. 2007, Pages 1-3.