Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
CAS: 142-31-4 | C8H15Na·H2SO4 | 232.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR416501000
Price (MYR)
12,862.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
12,862.00
Add to cart
Specifications
Chemical Name or MaterialOctyl sulfate, sodium salt
CAS142-31-4
Absorbance(5% aq. soln.) (1cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.02 max. at 250nm
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye damage.
View more
for ion pair chromatography
RUO – Research Use Only