Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
CAS: 142-31-4 | C8H15Na·H2SO4 | 232.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR416500250
Price (MYR)
4,460.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,460.00
Add to cart
Chemical Identifiers
CAS32287-65-3
IUPAC Nametrifluoroalumane hydrate
Molecular FormulaAlF3H2O
InChI KeyXTXFDUABTPNTFB-UHFFFAOYSA-K
SMILESO.F[Al](F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
for ion pair chromatography
RUO – Research Use Only