4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
CAS: 104-04-1 | C8H8N2O3 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR415751000
Price (MYR)
2,234.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,234.00
Add to cart
Chemical Identifiers
CAS12125-01-8
IUPAC Nameamine dihydrofluoride
Molecular FormulaF2H5N
InChI KeyKVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILESN.F.F
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.01%
Trace Metals(as Pb) 5ppm
Impurity contentSO₄ 0.005%
Assay (unspecified)98.0% NH₄F min
Impurity contentCl 0.001%