4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
CAS: 29270-56-2 | C6H2FN3O3 | 183.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
Catalog number ACR415450050
Price (MYR)
1,111.00
Add to cart
Quantity:
5 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,111.00
Add to cart
Chemical Identifiers
CAS618-95-1
IUPAC Namemethyl 3-nitrobenzoate
Molecular FormulaC8H7NO4
InChI KeyAXLYJLKKPUICKV-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (GC)≥98.0%
FormCrystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)76-80?C