α-Naphtholbenzein
α-Naphtholbenzein
α-Naphtholbenzein
Thermo Scientific Chemicals
α-Naphtholbenzein
Indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ACR415321000
Price (MYR)
2,580.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,580.00
Add to cart
Chemical Identifiers
CAS622-40-2
IUPAC Name2-(morpholin-4-yl)ethan-1-ol
Molecular FormulaC6H13NO2
InChI KeyKKFDCBRMNNSAAW-UHFFFAOYSA-N
SMILESOCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Water=<0.5 %
Infrared spectrumConforms
Refractive index1.4760 to 1.4780 (20°C, 589 nm)
Appearance (Form)Liquid